Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSBG2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526853

Catalog No. PIPA526853

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ACSBG2
Polyclonal
PBS with 0.09% sodium azide
Q5FVE4, Q2XU92
BGR, BRGL, PRTD-NY3, PRTDNY3, bubblegum-related protein, long-chain-fatty-acid--CoA ligase ACSBG2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
328845, 81616
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ACSBG2
Liquid
ACSBG2
KLH conjugated synthetic peptide between 237-265 amino acids from the central region of human ACBG2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit