Learn More
ASIC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00101W
Catalog No. PIOSR00101W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Probable cation channel with high affinity for sodium. These channels have been implicated in synaptic transmission, pain perception as well as mechanoperception. This gene is predomitly expressed in the pituitary gland. Expressed in brain, spinal chord and dorsal root ganglion (DRG). Expressed by a subset of sensory neurons in the DRG. Expressed by granule cells in the cerebellar cortex. In hippocampus, expression is detected in dentate gyrus granule cells, in pyramidal cells of CA1-CA3 subfields and in interneurons of the striatum oriens and radiatum of all subfields. In cerebral cortex expressed in small, medium and large pyramidal cells in layers 2, 3 and 5 respectively. Also expressed in striatum, globus pallidus, inferior and superior calliculi, amygdala, magnocellular preoptic nucleus, islands of Calleja and large neurons of olfactory tubercules. Highly expressed in newborn spinal chord but hardly detected in the cerebellum compared to adult. Expressed at postnatal day 1 in ependymal cells lining the central canal of spinal chord and in motor neurons. In adult, expression decreases in ependymal cells and increases in motor neurons. The number of positive interneurons decreases but the individual interneuron expression increases in adult spinal chord compared to newborn. In vitro, has no proton-gated channel activity.
Specifications
Specifications
ASIC4
Polyclonal
whole serum with no preservative
Q96FT7, Q7TNS7, Q9JHS6
Amiloride-sensitive cation channel 4 pituitary, acid-sensing ion channel 4, spinal chord ASIC, Spasic, BNAC4, MGC17248, MGC24860
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
ASIC4
Lyophilized
ASIC4
A synthetic peptide from internal region of human amiloride sensitive cation channel 4
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
241118, 55515, 63882