Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-Histone H4 Rabbit anti-Tetrahymena, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06598
Description
Anti-acetyl-Histone H4 Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a and has been validated in Chimmunoprecipitation, immunocytochemistry, immunoprecipitation and western blotting.Specifications
acetyl-Histone H4 | |
Polyclonal | |
Unconjugated | |
HIST1H4A; H4/J; HIST1H4F; HIST1H4L; H4FN; H4FA; H4FH; HIST1H4H; H4F2; H4FM; H4/A; H4FK; H4FG; HIST2H4; H4FB; H4/K; HIST1H4I; H4/N; HIST1H4B; H4FE; H4/M; H4/a; HIST1H4E; HIST4H4; HIST2H4A; H4FC; H4FI; H4/H; HIST1H4C; H4/G; HIST1H4K; H4FJ; H4FD; H4/I; H4/B; H4/D; H4/C; HIST1H4J; HIST1H4D; H4/E | |
KLH-conjugated peptide corresponding to amino acids 2-19 of Tetrahymena histone H4 (AGG(Ac)KGG(Ac)KGMG(Ac)KVGA(Ac)KRHS-C), acetylated on lysines 5, 8, 12 and 16. | |
200 μg | |
NM_175054 | |
−20°C for 12 months. Avoid repeated freeze/thaw cycles | |
IgG |
ChIP Assay, Immunocytochemistry, Immunoprecipitation, Western Blot | |
1mg/mL | |
Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide. | |
Rabbit | |
Protein A chromatography | |
Primary | |
Human, Tetrahymena species | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction