Learn More
Acetyl Histone H4 (Lys16) Mouse anti-Human, Clone: 4E10.2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051232
Catalog No. 05-123-2MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-acetyl Histone H4 (Lys16) Antibody, clone 4E10.2 is a mouse monoclonal antibody for detection of acetyl Histone H4 (Lys16) also known as H4K16Ac, Histone H4 (acetyl K16), HIST1H4 and has been validated in western blot, PIA.
Specifications
Specifications
acetyl Histone H4 (Lys16)
4E10.2
Purified from ascites by protein G chromatography. Supplied at 1mg/mL in PBS, 0.05% sodium azide.
Purified
KLH-conjugated, synthetic peptide containing the sequence (GKGGA[acetyl-K]RHRKC, corresponding to the sequence surrounding acetylated lysine 16 of human histone H4.
Protein G purfied
2-8°C for one year
Monoclonal
Inhibition Assays, Western Blot
Unconjugated
NP_003529
Murine
IgG1, κ
100μg
Primary
Human