Learn More
Acetyl (Lys26) phospho (Ser27) Histone H1.4 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061372
Catalog No. 06-137-2MI
Add to cart
Description
Description
Anti-acetyl (Lys26) phospho (Ser27) Histone H1.4 Antibody is a rabbit polyclonal antibody for detection of acetyl (Lys26) phospho (Ser27) Histone H1.4 also known as Histone H1.4, Histone H1b and has been validated in western blot, DB.
Specifications
Specifications
acetyl (Lys26) phospho (Ser27) Histone H1.4
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Dot Blot, Western Blot
Purified rabbit polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
HIST1H1E; H1F4
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H1.4 acetylated at Lys26 and phosphorylated at Ser27.
100μL
Primary
NP_005312