Learn More
Acetyl Lysine Mouse anti-All, Clone: 4G12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 16272
Description
Detect acetyl Lysine with Anti-acetyl Lysine Antibody, clone 4G12, agarose conjugate (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation.
Specifications
acetyl Lysine | |
Monoclonal | |
Agarose | |
A mixture of chemically acetylated antigens. Clone 4G12. | |
Primary | |
4°C for one year. Do not freeze. |
Immunoprecipitation | |
4G12 | |
Mouse | |
100 μg | |
All species |