Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylcholinesterase Goat anti-Bovine, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA126888

Catalog No. PIPA126888


Add to cart

Description

Description

PA1-26888 detects Acetylcholinesterase from bovine samples. PA1-26888 has been successfully used in Western blot, ELISA, and immunoprecipitation procedures. The PA1-26888 immunogen is Acetyl Cholinesterase from bovine erythrocytes. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Acetylcholinesterase
Polyclonal
Unconjugated
ACHE
Liquid
Goat
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
540446
ELISA, Immunoprecipitation, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
P23795
ACHE
Acetyl Cholinesterase from bovine erythrocytes.
IgG fraction
RUO
Antibody
Polyclonal
Bovine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit