Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylcholinesterase Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521371

Catalog No. PIPA521371

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA5-21371 targets AChE in ICC, IF, and WB applications and shows reactivity with Human samples. The PA5-21371 immunogen is a recombit fragment corresponding to a region within amino acids 406 and 614 of AChE (Uniprot IDNo.P22303). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Acetylcholinesterase
Polyclonal
Unconjugated
ACHE
Liquid
ACHE
Recombit fragment corresponding to a region within amino acids 406 and 614 of AChE (Uniprot IDNo.P22303)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.73 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
P22303, P21836, P37136
ACEE, ARACHE, N-ACHE, YT, Yt blood group, apoptosis-related acetylcholinesterase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11423, 43, 83817
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit