Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Phosphatase 1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141704

Catalog No. 01-675-109


Add to cart

Description

Description

Suggested positive control: antigen standard for ACP1 (transient overexpression lysate), human liver protein, liver lysate.

ACP1 is a unique member of Phosphotyrosine Protein phosphotase family of proteins. It is ubiquitously expressed acid phosphotase functioning as an acid phosphotase and a protein tyrosine phosphotase. ACP1 has a cysteine-based catalytic mechanism and hydrolyzes protein tyrosine phosphate to protein tyrosine and orthophosphate. Reports suggest a functional role of ACP1 in TCR signaling and control of cell-cell adhesion through regulation of cytoskeletion assembly.
Specifications

Specifications

Acid Phosphatase 1
Polyclonal
Unconjugated
ACP1
Liquid
ACP1
Synthetic peptide corresponding to residues 50-100 of human ACP1 .
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
P24666
ACP1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
52
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit