Learn More
ACK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA528389
Catalog No. PIPA528389
Add to cart
Description
Description
PA5-28390 targets ERR alpha in WB applications and shows reactivity with Human samples. The PA5-28390 immunogen is recombit fragment corresponding to a region within amino acids 69 and 359 of Human ERR alpha.
This gene encodes a tyrosine kinase that binds Cdc42Hs in its GTP-bound form and inhibits both the intrinsic and GTPase-activating protein (GAP)-stimulated GTPase activity of Cdc42Hs. This binding is mediated by a unique sequence of 47 amino acids C-terminal to an SH3 domain. The protein may be involved in a regulatory mechanism that sustains the GTP-bound active form of Cdc42Hs and which is directly linked to a tyrosine phosphorylation signal transduction pathway. Several alternatively spliced transcript variants have been identified from this gene, but the full-length nature of only two transcript variants has been determined.
Specifications
Specifications
ACK1
Polyclonal
Unconjugated
TNK2
Liquid
TNK2
Recombit fragment corresponding to a region within amino acids 25 and 313 of Human ACK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q07912, O54967
ACK, ACK-1, ACK1, p21cdc42Hs, activated CDC42 kinase 1, activated Cdc42-associated kinase 1, activated p21cdc42Hs kinase, tyrosine kinase non-receptor protein 2
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10188, 51789