Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aconitase 1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527824

Catalog No. PIPA527824


Add to cart

Description

Description

PA5-27824 targets Aconitase 1 in WB applications and shows reactivity with Human samples. The PA5-27824 immunogen is recombit fragment corresponding to a region within amino acids 83 and 461 of Human Aconitase 1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5 ft. UTR of ferritin mRNA, and in the 3 ft. UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes.
Specifications

Specifications

Aconitase 1
Polyclonal
Unconjugated
ACO1
Liquid
ACO1
Recombit fragment corresponding to a region within amino acids 83 and 461 of Human Aconitase 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.78 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P21399
ACONS, IREB1, IREBP, IREBP1, IRP1, IRE-BP 1, aconitate hydratase, citrate hydro-lyase, cytoplasmic aconitate hydratase, ferritin repressor protein, iron regulatory protein 1, iron-responsive element binding protein 1, iron-responsive element-binding protein 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
48
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit