Learn More
ACOT11 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523639
Catalog No. PIPA523639
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene encodes a member of the acyl-CoA thioesterase family which catalyse the conversion of activated fatty acids to the corresponding non-esterified fatty acid and coenzyme A. Expression of a mouse homolog in brown adipose tissue is induced by low temperatures and repressed by warm temperatures. Higher levels of expression of the mouse homolog has been found in obesity-resistant mice compared with obesity-prone mice, suggesting a role of acyl-CoA thioesterase 11 in obesity. Alternative splicing results in transcript variants.
Specifications
Specifications
ACOT11
Polyclonal
Unconjugated
ACOT11
Liquid
ACOT11
KLH conjugated synthetic peptide between 549-575 amino acids from the C-terminal region of human ACOT11
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8WXI4
BFIT, STARD14, THEA, THEM1, START domain containing 14, StAR-related lipid transfer (START) domain containing 14, acyl-CoA thioester hydrolase 11, acyl-coenzyme A thioesterase 11, adipose-associated thioesterase, brown fat-inducible thioesterase, thioesterase superfamily member 1, thioeste
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26027