Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACOT8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132452
Description
ACOT8 Polyclonal specifically detects ACOT8 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ACOT8 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
ACOT8 | |
Recombinant protein encompassing a sequence within the center region of human ACOT8. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
10005 | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
1.0mg/mL | |
Western Blot 1:500-1:3000 | |
ACTEIII, acyl-CoA thioesterase 8HNAACTE, Choloyl-coenzyme A thioesterase, EC 3.1.2.27, hACTEIII, HIV-Nef-associated acyl-CoA thioesterase, hTEcholoyl-CoA hydrolase, long-chain fatty-acyl-CoA hydrolase, palmitoyl-CoA hydrolase, peroxisomal acyl-CoA thioesterase, peroxisomal acyl-CoA thioesterase 1, Peroxisomal acyl-coenzyme A thioester hydrolase 1, Peroxisomal long-chain acyl-CoA thioesterase 1, PTE-1, PTE1HIV-Nef associated acyl-CoA thioesterase, PTE2, PTE-2hACTE-IIIacyl-coenzyme A thioesterase 8, Thioesterase II, thioesterase III | |
Rabbit | |
36 kDa | |
100 μL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction