Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

acyl-Coenzyme A oxidase 2, branched chain, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ACOX2 protein.

Manufacturer:  Abnova Corporation H00008309B01

Catalog No. 89-006-552


Add to cart

Description

Description

The product of this gene belongs to the acyl-CoA oxidase family. It encodes the branched-chain acyl-CoA oxidase which is involved in the degradation of long branched fatty acids and bile acid intermediates in peroxisomes. Deficiency of this enzyme results in the accumulation of branched fatty acids and bile acid intermediates, and may lead to Zellweger syndrome, severe mental retardation, and death in children. [provided by RefSeq

Sequence: MGSPVHRVSLGDTWSRQMHPDIESERYMQSFDVERLTNILDGGAQNTALRRKVESIIHSYPEFSCKDNYFMTQNERYKAAMRRAFHIRLIARRLGWLEDGRELGYAYRALSGDVALNIHRVFVRALRSLGSEEQIAKWDPLCKNIQIIATYAQTELGHGTYLQGLETEATYDAATQEFVIHSPTLTATKWWPGDLGRSATHALVQAQLICSGARRGMHAFIVPIRSLQDHTPLPGIIIGDIGPKMDFDQTDNGFLQLNHVRVPRENMLSRFAQVLPDGTYVKLGTAQSNYLPMVVVRVELLSGEILPILQKACVIAMRYSVIRRQSRLRPSDPEAKVLDYQTQQQKLFPQLAISYAFHFLAVSLLEFFQHSYTAILNQDFSFLPELHALSTGMKAMMSEFCTQGAEMCRRACGGHGYSKLSGLPSLVTKLSASCTYEGENTVLYLQVARFLVKSYLQTQMSPGSTPQRSLSPSVAYLTAPDLARCPAQRAADFLCPELYTTAWAHVAVRLIKDSVQHLQTLTQSGADQHEAWNQTTVIHLQAAKVHCYYVTVKGFTEALEKLENEPAIQQVLKRLCDLHAIHGILTNSGDFLHDAFLSGAQVDMARTAYLDLLRLIRKDAILLTDAFDFTDQCLNSALGCYDGNVYERLFQWAQKSPTNTQENPAYEEYIRPLLQSWRSKL
Specifications

Specifications

acyl-Coenzyme A oxidase 2, branched chain
Unconjugated
No additive
NM_003500.2
BCOX|BRCACOX|BRCOX|THCCox
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
8309
Western Blot
Mouse polyclonal antibody raised against a full-length human ACOX2 protein.
ACOX2
Serum
ACOX2
ACOX2 (NP_003491.1, 1 a.a. ~ 681 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit