Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRACP/PAP/ACP5 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131535

Catalog No. NBP131535


Add to cart

Description

Description

TRACP/PAP/ACP5 Polyclonal antibody specifically detects TRACP/PAP/ACP5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

TRACP/PAP/ACP5
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
ACP5
Recombinant protein encompassing a sequence within the center region of human TRAP. The exact sequence is proprietary.
37 kDa
100 ul
Protein Phosphatase
Primary
54
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
acid phosphatase 5, tartrate resistant, EC 3.1.3.2, MGC117378, Tartrate-resistant acid ATPase, tartrate-resistant acid phosphatase type 5, TRAP, TR-AP, TrATPase, Type 5 acid phosphatase
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only