Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-ACP6, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals 45380002
Description
ACP6 Polyclonal antibody specifically detects ACP6 in Human samples. It is validated for Western Blot, ELISA.Specifications
ACP6 | |
Unconjugated | |
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative | |
Affinity Purified | |
ACP6 | |
This antibody is specific for the Middle Region of the target protein. | |
Immunogen affinity purified | |
RUO | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
This product is specific for Human ACP6. |
ELISA, Western Blot | |
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000 | |
Q9NPH0 | |
acid phosphatase 6, lysophosphatidicPACPL1, Acid phosphatase-like protein 1, ACPL1acid phosphatase like 1, EC 3.1.3.2, LPAPlysophosphatidic acid phosphatase type 6 | |
Rabbit | |
IgG | |
0.05 mg | |
Lipid and Metabolism | |
Primary | |
51205 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only