Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostate Specific Acid Phosphatase Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514545

Catalog No. PIPA514545


Add to cart

Description

Description

ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.
Specifications

Specifications

Prostate Specific Acid Phosphatase
Polyclonal
Unconjugated
ACPP
Liquid
ACPP
KLH conjugated synthetic peptide between 313-341 amino acids from the C-terminal region of human ACPP
Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P15309, Q8CE08
Prostatic acid phosphatase [Precursor]; PPAP; EC 3.1.3.2; ACPP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55, 56318
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit