Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrp30/Adiponectin Mouse anti-Human, Unlabeled, Clone: 31, BD

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 611644

 View more versions of this product

Catalog No. BDB611644

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.


Description

Description

Adipocytes express a variety of proteins that function in the homeostatic control of glucose and lipid metabolism. Insulin regulates the translocation and secretion of many of these adipocyte proteins in response to homeostatic changes in energy balance. Both adipsin and adipocyte complement-related protein of 30kDa (Acrp30) are secretory proteins whose secretion from adipocytes is enhanced by insulin stimulation. Acrp30 contains N-terminal collagen repeats and a globular C-terminal domain. It is homologous to collagen type VIII and X and to complement factor C1q. Also, it forms homo-oligomeric structures of 90 and 300kDa. The expression of Acrp30 increases 100 fold during adipocyte differentiation and it is commonly found in serum. Insulin stimulation of 3T3-L1 fibroblasts leads to a 4-fold increase in Acrp30 secretion. This secretion is regulated by PI3-kinase activity and may involve a secretory pathway that is distinct from other insulin-regulated secretory pathways, such as that involved with the translocation of glucose transporters and transferrin receptors. Thus, Acrp30 is an adipocyte secretory protein that functions in the regulation of energy homeostasis.

Host Species: Mouse
Clone: 31
Isotype: IgG2a
Species Reactivity [for Features Main]: Human
Immunogen: Human Acrp30 aa. 1-247

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Acrp30/Adiponectin
Monoclonal
250μg/mL
Adiponectin
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
31
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Acrp30 aa. 1-247
50 μg
Cell Biology
Human
IgG2a
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.