Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrp30/Adiponectin Mouse anti-Human, Unlabeled, Clone: 31, BD

Mouse Monoclonal Antibody

$207.00

Specifications

Antigen Acrp30/Adiponectin
Isotype IgG2a
Immunogen Human Acrp30 aa. 1-247
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB611644
View Documents
BD Biosciences
611644
50μg
Each for $207.00
Add to cart
 
BDB611645
View Documents
BD Biosciences
611645
150μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Adipocytes express a variety of proteins that function in the homeostatic control of glucose and lipid metabolism. Insulin regulates the translocation and secretion of many of these adipocyte proteins in response to homeostatic changes in energy balance. Both adipsin and adipocyte complement-related protein of 30kDa (Acrp30) are secretory proteins whose secretion from adipocytes is enhanced by insulin stimulation. Acrp30 contains N-terminal collagen repeats and a globular C-terminal domain. It is homologous to collagen type VIII and X and to complement factor C1q. Also, it forms homo-oligomeric structures of 90 and 300kDa. The expression of Acrp30 increases 100 fold during adipocyte differentiation and it is commonly found in serum. Insulin stimulation of 3T3-L1 fibroblasts leads to a 4-fold increase in Acrp30 secretion. This secretion is regulated by PI3-kinase activity and may involve a secretory pathway that is distinct from other insulin-regulated secretory pathways, such as that involved with the translocation of glucose transporters and transferrin receptors. Thus, Acrp30 is an adipocyte secretory protein that functions in the regulation of energy homeostasis.

Host Species: Mouse
Clone: 31
Isotype: IgG2a
Species Reactivity [for Features Main]: Human
Immunogen: Human Acrp30 aa. 1-247

Immunofluorescence, Western Blotting

Specifications

Specifications

Acrp30/Adiponectin
Human Acrp30 aa. 1-247
Unconjugated
Cell Biology
Murine
Affinity Purified
Monoclonal
Adiponectin
IgG2a
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
31
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit