Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acrosomal Vesicle Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA544074
Description
Peptide sequence: MNRFLLLMSL YLLGSARGTS SQPNELSGSI DHQTSVQQLP GEFFSLENPS Sequence homology: Guinea Pig: 79%; Human: 100%; Rabbit: 85%
Acrosomal Vesicle Protein Polyclonal antibody specifically detects Acrosomal Vesicle Protein in Human samples. It is validated for Western Blot.Specifications
Acrosomal Vesicle Protein | |
Polyclonal | |
Unconjugated | |
ACRV1 | |
acrosomal protein SP-10; Acrosomal vesicle protein 1; Acrv1; D11S4365; Msa63; MSA-63; rSP10; Sp10; SP-10; SPACA2; sperm membrane protein; sperm protein 10 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P26436 | |
ACRV1 | |
Synthetic peptide directed towards the N-terminal of human ACRV1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction