Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSF3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525803

Catalog No. PIPA525803


Add to cart

Specifications

Specifications

ACSF3
Polyclonal
PBS with 0.09% sodium azide
Q4G176, Q3URE1
acyl-CoA synthetase family member 3, mitochondrial
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
197322, 257633
Immunohistochemistry, Western Blot
Unconjugated
ACSF3
Liquid
ACSF3
KLH conjugated synthetic peptide between 413-442 amino acids from the central region of human ACSF3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit