Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSM1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526292

Catalog No. PIPA526292

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ACSM1 has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). This protein acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy-and 2,3-or 3,4-unsaturated acids (in vitro). This protein functions as GTP-dependent lipoate-activating enzyme that generates the substrate for lipoyltransferase.
Specifications

Specifications

ACSM1
Polyclonal
PBS with 0.09% sodium azide
Q08AH1
BUCS1, MACS1, Butyrate CoA ligase, acyl-coenzyme A synthetase ACSM1, mitochondrial, butyrate--CoA ligase 1, butyryl Coenzyme A synthetase 1, butyryl-coenzyme A synthetase 1, lipoate-activating enzyme, medium-chain acyl-CoA synthetase, middle-chain acyl-CoA synthetase 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
116285
Flow Cytometry, Western Blot
Unconjugated
ACSM1
Liquid
ACSM1
KLH conjugated synthetic peptide between 21-48 amino acids from the N-terminal region of human ACSM1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit