Learn More
POMC Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136035
Catalog No. 01-672-225
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
ATCH (adrenocorticotropic hormone) is a hormone which plays a major role in stimulating the adrenal cortex. It is formed through cleavage of the polypeptide precursor proopiomelanocortin (POMC), which also results in several other cleavage products including MSH, ACTH, and beta endorphin. ATCH is secreted from the anterior pituitary in response to the corticotropin-releasing hormone from the hypothalamus. It stimulates the secretion of glucocorticoids like cortisol, but has little control over the stimulation of mineralocorticoids like aldosterone, which is another major hormone of the adrenal cortex.
Specifications
Specifications
POMC
Polyclonal
whole serum with no preservative
P01189, P01193, P01192, P01194
Adrenocorticotropic Hormone, Corticotropin
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Porcine, Rat
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
POMC
Liquid
POMC
Synthetic ACTH.
250 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18976, 24664, 396863, 5443