Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POMC Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137013

Catalog No. 01-673-018


Add to cart

Description

Description

A suggested positive control is normal pituitary gland or pituitary adenoma.

Human adrenocorticotrophin (ACTH) is a 39 amino acid cleavage product from pro-opiomelanocortin (POMC) which elicits a classic glucocorticoid response from the adrenal glands in situations of stress. ACTH is produced in the pituitary, and also by lymphocytes and macrophages.
Specifications

Specifications

POMC
Polyclonal
PBS with 1% BSA and <0.1% sodium azide
P01189
Adrenocorticotrophic Hormone
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5443
Immunohistochemistry (Paraffin)
Unconjugated
POMC
Liquid
POMC
Synthetic peptide derived from N-terminus of human ACTH
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit