Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Actin Alpha 2 Smooth Muscle, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP135269

Catalog No. NBP135269

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

alpha-Smooth Muscle Actin
Unconjugated
TBS (pH 7.6) and 1.0% BSA with 0.1% Sodium Azide
AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
This product is specific for Actin-Smooth Muscle
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin Neat
Affinity Purified
ACTA2
A synthetic peptide from N-terminus of human alpha smooth muscle isoform of actin
42 kDa
7 ml
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Stem Cell Markers
Primary
59
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only