Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Skeletal Muscle Actin Mouse anti-Human, Clone: 5C5.F8.C7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA137021

Catalog No. 01-671-632


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is skeletal muscle tissue.

MA1-37021 is highly specific and shows no cross-reaction with smooth muscle actin. This antibody reacts with sarcomeric actins of normal tissues and neoplasms derived from such tissues (i.e. rhabdomyosarcomas).
Specifications

Specifications

Skeletal Muscle Actin
Monoclonal
Unconjugated
ACTA1
Liquid
ACTA1
N-terminus decapeptide of alpha skeletal muscle isoform of actin
Protein A/G
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
5C5.F8.C7
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P68133
ACTA1
Mouse
IgM
100 μL
4° C, do not freeze
Primary
58
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit