Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Skeletal Muscle Actin Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137020

Catalog No. 01-673-021


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, amphibian, mouse, porcine, rabbit and rat based on sequence homology. A suggested positive control is skeletal muscle tissue.

This is highly specific and shows no cross-reaction with smooth muscle actin. This antibody reacts with sarcomeric actins of normal tissues and neoplasms derived from such tissues (i.e. rhabdomyosarcomas).
Specifications

Specifications

Skeletal Muscle Actin
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
P68133, P68134, P62736, P68136
ACTA1
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
59, 11459, 58, 29437
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ACTA1
Liquid
Acta1, ACTA2
Synthetic peptide derived from N-terminus of human alpha skeletal muscle isoform of actin
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit