Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Activin A beta A subunit Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 9 publications
Supplier: R&D Systems MAB3381500
This item is not returnable.
View return policy
Description
Activin A Monoclonal specifically detects Activin A in Human, Mouse, Rat samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
Activin A | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
INHBA | |
Chinese hamster ovary cell line CHO-derived recombinant human Activin A | |
500 μg | |
Primary | |
Detects recombinant human Activin A in ELISAs. Also recognizes Activin A precursor. The amino acid sequences of mature beta A subunits from human, mouse and rat are identical. Therefore, this antibody will also detect the mature beta A subunit from mouse and rat. | |
Human, Mouse, Rat | |
Supernatant |
Neutralization, ELISA | |
69403 | |
Neutralization 0.02-0.06 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
P18075 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
655 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction