Learn More
Activin A βA subunit Mouse anti-Human, Mouse, Rat, Clone: 69403, R&D Systems™
Mouse Monoclonal Antibody has been used in 9 publications
Manufacturer:  R&D Systems MAB3381SP
 View more versions of this product
Catalog No. MAB3381SP
Add to cart
This item is not returnable. View return policy
Description
Description
Activin A Monoclonal specifically detects Activin A in Human, Mouse, Rat samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
Activin A
69403
Neutralization 0.02-0.06 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Purified
Mouse
IgG1
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects recombinant human Activin A in ELISAs. Also recognizes Activin A precursor. The amino acid sequences of mature beta A subunits from human, mouse and rat are identical. Therefore, this antibody will also detect the mature beta A subunit from mouse and rat.
Human, Mouse, Rat
Neutralization, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
INHBA
Chinese hamster ovary cell line CHO-derived recombinant human Activin A
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3624
Reconstitute at 0.5 mg/mL in sterile PBS.