Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin A βA subunit Mouse anti-Human, Mouse, Rat, Clone: 69403, R&D Systems™

Mouse Monoclonal Antibody

$135.50 - $839.00

Specifications

Antigen Activin A
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Activin A
Clone 69403
Applications Neutralization, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB3381SP
View Documents
R&D Systems
MAB3381SP
25 ug
Each for $135.50
Add to cart
 
MAB3381100
View Documents
R&D Systems
MAB3381100
100 ug
Each for $377.00
Add to cart
 
MAB3381500
View Documents
R&D Systems
MAB3381500
500 ug
Each for $839.00
Add to cart
 
Description

Description

Activin A βA subunit Monoclonal antibody MAB3381 Specifically detects Activin A βA subunit in Human, Mouse, Rat samples. It is validated for ELISA Capture, Neutralization
Specifications

Specifications

Activin A
69403
Unconjugated
Mouse
IgG1
3624
RUO
Chinese hamster ovary cell line CHO-derived recombinant human Activin A
Neutralization, ELISA
Purified
Monoclonal
Primary
Detects recombinant human Activin A in ELISAs. Also recognizes Activin A precursor. The amino acid sequences of mature beta A subunits from human, mouse and rat are identical. Therefore, this antibody will also detect the mature beta A subunit from mouse and rat.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit