Learn More
anti-Activin RIA/ALK-2/Activin Receptor Type 1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP133096
Catalog No. NBP133096
Add to cart
Description
Description
Activin RIA/ALK-2/Activin Receptor Type 1 Polyclonal specifically detects Activin RIA/ALK-2/Activin Receptor Type 1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Activin RIA/ALK-2/Activin Receptor Type 1
0.41mg/mL
Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
ACVR1
Recombinant protein encompassing a sequence within the center region of human Activin A Receptor type I. The exact sequence is proprietary.
57 kDa
0.1 ml
Protein Kinase
Primary
90
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
activin A receptor, type I, ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human.