Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Activin RIA/ALK-2/Activin Receptor Type 1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133500

Catalog No. NBP133500


Add to cart

Description

Description

Activin RIA/ALK-2/Activin Receptor Type 1 Polyclonal antibody specifically detects Activin RIA/ALK-2/Activin Receptor Type 1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

Activin RIA/ALK-2/Activin Receptor Type 1
0.9mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000
Affinity Purified
ACVR1
Recombinant protein encompassing a sequence within the center region of human Activin A Receptor type I. The exact sequence is proprietary.
57 kDa
0.1 ml
Protein Kinase
Primary
90
Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
activin A receptor, type I, ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only