Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAF53B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525247

Catalog No. PIPA525247

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ACTL6B is a member of a family of actin-related proteins (ARPs) which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene encodes a subunit of the BAF (BRG1/brm-associated factor) complex in mammals, which is functionally related to SWI/SNF complex in S. cerevisiae and Drosophila; the latter is thought to facilitate transcriptional activation of specific genes by antagonizing chromatin-mediated transcriptional repression. This subunit may be involved in the regulation of genes by structural modulation of their chromatin, specifically in the brain.
Specifications

Specifications

BAF53B
Polyclonal
PBS with 0.09% sodium azide
O94805
ACTL6, BAF53B, 53 kDa BRG1-associated factor B, BRG1-associated factor 53B, actin-like 6, actin-like protein 6B, actin-related protein Baf53b, arpNalpha, hArpN alpha
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51412
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ACTL6B
Liquid
ACTL6B
KLH conjugated synthetic peptide between 42-70 amino acids from the N-terminal region of human ACTL6B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit