Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR1A Goat anti-Human, Mouse, Porcine, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131098

Catalog No. PIPA131098


Add to cart

Description

Description

PA1-31098 detects ACTR1A from human samples. PA1-31098 has been successfully used in Western blot applications. The PA1-31098 immunogen is: Synthetic peptide: YEEDGARSIHRKT (Human) (C terminal). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a 42.6 kD subunit of dynactin, a macromolecular complex consisting of 10-11 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein. It is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit is present in 8-13 copies per dynactin molecule, and is the most abundant molecule in the dynactin complex. It is an actin-related protein, and is approximately 60% identical at the amino acid level to conventional actin.
Specifications

Specifications

ACTR1A
Polyclonal
Unconjugated
ACTR1A
Liquid
ACTR1A
synthetic peptide YEEDGARSIHRKT (Human) (C terminal).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Porcine, Rat
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P61163, P61164, P85515
ARP1, CTRN1, FLJ52695, FLJ52800, FLJ55002
Goat
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100156619, 10121, 294010, 54130
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit