Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTR1B Mouse anti-Human, Clone: 4E10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA520934

Catalog No. PIMA520934


Add to cart

Description

Description

Peptide Sequence: SRYLRLLLRK EGVDFHTSAE FEVVRTIKER ACYLSINPQK DEALETEKVQ YTLPDGSTLD VGPARFRAPE LLFQPDLVGD ESEGLHEVVA FAIHK*

This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex.
Specifications

Specifications

ACTR1B
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P42025
ACTR1B; ARP1B; CTRN2; ARP1 actin-related protein 1 homolog B, centractin beta (yeast); PC3; centractin beta; ARP1, yeast homolog B; ARP1 (actin-related protein 1, yeast) homolog B (centractin beta); ARP1 actin-related protein 1 homolog B, centractin beta
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10120
ELISA, Immunocytochemistry, Immunofluorescence
4E10
Unconjugated
ACTR1B
Liquid
ACTR1B
ACTR1B (NP_005726, 191 a.a. ∽ 286 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit