Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARP2 actin-related protein 2 homolog (yeast), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ACTR2 protein.

Manufacturer:  Abnova Corporation H00010097B01

Catalog No. 89-006-893


Add to cart

Description

Description

The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MVGDEASELRSMLEVNYPMENGIVRNWDDMKHLWDYTFGPEKLNIDTRNCKILLTEPPMNPTKNREKIVEVMFETYQFSGVYVAIQAVLTLYAQGLLTGVVVDSGDGVTHICPVYEGFSLPHLTRRLDIAGRDITRYLIKLLLLRGYAFNHSADFETVRMIKEKLCYVGYNIEQEQKLALETTVLVESYTLPDGRIIKVGGERFEAPEALFQPHLINVEGVGVAELLFNTIQAADIDTRSEFYKHIVLSGGSTMYPGLPSRLERELKQLYLERVLKGDVEKLSKFKISIEDPPRRKHMVFLGGAVLADIMKDKDNFWMTRQEYQEKGVRVLEKLGVTVR
Specifications

Specifications

ARP2 actin-related protein 2 homolog (yeast)
Unconjugated
No additive
BC036253.1
ARP2
Mouse
50 uL
Kinases
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human ACTR2 protein.
ACTR2
Serum
ACTR2
ACTR2 (AAH36253.1, 1 a.a. ~ 339 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
10097
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit