Learn More
Ad6, Mouse, Clone: 143, Abnova™
Mouse monoclonal antibody raised against adenovirus type 6.
Manufacturer:  Abnova Corporation MAB3882
Catalog No. 89-024-824
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Ad6
143
Mouse monoclonal antibody raised against adenovirus type 6.
In 100mM PBS, pH 7.2
Murine
IgG1
RUO
Primary
Specific for the hexon group antigen of many Adenovirus serotypes. Known reactivity with 34 serotypes of Adenovirus including types 40 and 41 (40, 41, 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 14, 16, 18, 19, 20, 26, 31, 34, 35, 36 and 37). Does not react with Influenza A, Influenza B, RSV, Parainfluenza 1, 2 & 3, Mycoplasma pneumonia, H. pylori and Mammalian cells.
ELISA, Immunofluorescence, Immunoprecipitation
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Adenovirus type 6 infected cells.
1 mg
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal