Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA125166

Catalog No. PIPA125166


Add to cart

Description

Description

PA1-25166 detects ADAM10 from Human, Rat, Mouse samples. PA1-25166 has been successfully used in Western blot procedures. Positive control: Jurkat whole cell lysates The PA1-25166 immunogen is a synthetic peptide corresponding to the cytoplasmic domain of human ADAM10. Store at 4°C short term. For extended storage aliquot and store at -20°C. Avoid freeze-thaw cycles. DO NOT store below -22°C.

Members of the ADAM family are cell surface proteins with a unique structure possessing both potential adhesion and protease domains. This gene encodes and ADAM family member that cleaves many proteins including TNF-alpha and E-cadherin.
Specifications

Specifications

ADAM10
Polyclonal
Unconjugated
ADAM10
Liquid
ADAM10
Synthetic peptide corresponding to the cytoplasmic domain of human ADAM10.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O14672, O35598, Q10743
AD10, HsT18717, MADM, kuz, CDw156
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
102, 11487, 29650
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit