Learn More
MilliporeSigma™ anti-ADAM12 Clone: 1G3, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1600100UG
Catalog No. ST1600100UG
Add to cart
Description
Description
Specifically detects ADAM12 Clone: 1G3 in Human samples, and it is validated for ELISA, Immuno Blotting
This gene encodes a member of the ADAM (a disintegrin and metalloprotease) protein family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene has two alternatively spliced transcripts: a shorter secreted form and a longer membrane-bound form. The shorter form is found to stimulate myogenesis.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ADAM12
1G3
In PBS, pH 7.2.
Murine
IgG3
RUO
Primary
Recognizes the ADAM12 protein.
ELISA, Immuno Blotting, Immunoblot
Unconjugated
Purified
A recombinant polypeptide corresponding to amino acids 208-305 of human ADAM12, expressed as a GST fusion protein
100μg
−20°C
Monoclonal
Human