Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM12, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA120301

Catalog No. PIPA120301

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

ADAM12 is a member of the ADAM (a disintegrin and metalloprotease-like domain) family. Two forms of ADAM12 have been described: ADAM12S and ADAM12L (short and long forms). The short form is a soluble form lacking the transmembrane and cytoplasmic domains. The short form of ADAM12 was reported to provoke myogenesis, and the lack of cytoplasmic domain suggests different regulation pathways (although both forms can be expressed in the same tissue).

PA1-20301 detects ADAM12 in human samples.PA1-20301 has been successfully used in Western blot procedures.The PA1-20301 immunogen is a synthetic peptide corresponding to the cytoplasmic domain of human ADAM12.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ADAM12
Polyclonal
Unconjugated
O43184
ADAM12
Synthetic peptide corresponding to the cytoplasmic domain of human ADAM12.
25 μg
Primary
Human
Antibody
IgG
Western Blot
1mg/mL
ADAM12
RP11-295J3.5, MCMP, MCMPMltna, MLTN, MLTNA
Rabbit
Antigen affinity chromatography
RUO
8038
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit