Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ADAM21, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232414

Catalog No. NBP232414


Add to cart

Description

Description

ADAM21 Polyclonal antibody specifically detects ADAM21 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ADAM21
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
a disintegrin and metalloproteinase domain 21, ADAM 21, ADAM metallopeptidase domain 21, ADAM metallopeptidase domain 21, preproprotein, ADAM31, disintegrin and metalloproteinase domain-containing protein 21, EC 3.4.24.-, MGC125389
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin 1:10 - 1:20
Affinity Purified
ADAM21
This antibody was developed against a recombinant protein corresponding to amino acids: MQHDEEFCFCGERGCIMNTFRVPAEKFTNCSYADFMKTTLNQGSCLHNPPRLGEIFMLKR
Immunogen affinity purified
RUO
Primary
8747
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only