Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM8 aa 158-497 Mouse anti-Human, Clone: 143303, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen ADAM8
Immunogen Mouse myeloma cell line NS0-derived recombinant human ADAM8 ectodomain Asp158-Ser653 Accession # P78325
Clone 143303
Applications Western Blot, Immunoprecipitation
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1031SP
View Documents
R&D Systems
MAB1031SP
25 ug
Each for $119.60
Add to cart
 
MAB1031100
View Documents
R&D Systems
MAB1031100
100 ug
Each for $377.00
Add to cart
 
MAB1031500
View Documents
R&D Systems
MAB1031500
500 ug
Each for $839.00
Add to cart
 
Description

Description

ADAM8 aa 158-497 Monoclonal antibody MAB1031 Specifically detects ADAM8 aa 158-497 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

ADAM8
143303
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2b
101
RUO
Mouse myeloma cell line NS0-derived recombinant human ADAM8 ectodomain Asp158-Ser653 Accession # P78325
Western Blot, Immunoprecipitation
Purified
P78325
Monoclonal
Primary
Detects human ADAM8 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with the ectodomains of recombinant human ADAM9, 10, 15, 17, and 19 is observed. Detects an epitope within aa 201-497 of recombinant human ADAM8 which corresponds to the metalloprotease and disintegrin-like domains.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit