Learn More
MilliporeSigma™ anti-ADAMTS1, Polyclonal Available on GSA/VA Contract for Federal Government customers only.
Mouse Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST1604100UG
Catalog No. ST1604100UG
Add to cart
Description
Description
Specifically detects ADAMTS1 Clone: 1B8 in Human samples, and it is validated for ELISA, Immuno Blotting
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene contains two disintegrin loops and three C-terminal TS motifs and has anti-angiogenic activity. The expression of this gene may be associated with various inflammatory processes as well as development of cancer cachexia. This gene is likely to be necessary for normal growth, fertility, and organ morphology and function.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ADAMTS1
1B8
In PBS, pH 7.27.
Murine
IgG2b
RUO
Primary
Recognizes the ADAMTS1 protein.
ELISA, Immuno Blotting, Immunoblot
Unconjugated
Purified
A recombinant polypeptide corresponding to amino acids 858-968 of human ADAMTS1, expressed as a GST fusion protein
100μg
−20°C
Polyclonal
Human