Learn More
ADAM metallopeptidase with thrombospondin type 1 motif, 13, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ADAMTS13.
Supplier: Abnova Corporation H00011093A01
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura. Alternative splicing of this gene generates multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: FINVAPHARIAIHALATNMGAGTEGANASYILIRDTHSLRTTAFHGQQVLYWESESSQAEMEFSEGFLKAQASLRGQYWTLQSWVPEMQDPQSWKGKEGTSpecifications
ADAM metallopeptidase with thrombospondin type 1 motif, 13 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant ADAMTS13. | |
ADAMTS13 | |
C9orf8/DKFZp434C2322/FLJ42993/MGC118899/MGC118900/TTP/VWFCP/vWF-CP | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_139025 | |
ADAMTS13 | |
ADAMTS13 (NP_620594, 1328 a.a. ∼ 1427 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
11093 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.