Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514339

Catalog No. PIPA514339

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ADAMTS13 is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme is the von Willebrand Factor (vWF)-cleaving protease, which is responsible for cleaving at the site of Tyr842-Met843 of the vWF molecule. A deficiency of this enzyme is associated with thrombotic thrombocytopenic purpura.
Specifications

Specifications

ADAMTS13
Polyclonal
PBS with 0.09% sodium azide
Q76LX8
von Willebrand factor-cleaving protease; vWF-cleaving protease; vWF-CP; A disintegrin and metalloproteinase with thrombospondin motifs 13; ADAMTS-13; ADAM-TS 13; ADAM-TS13; ADAMTS13; C9orf8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11093
Immunohistochemistry, Western Blot
Unconjugated
ADAMTS13
Liquid
ADAMTS13
KLH conjugated synthetic peptide between 829-858 amino acids from the central region of human ADAMTS13
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit