Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ADAMTS19, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP231627

Catalog No. NBP231627


Add to cart

Description

Description

ADAMTS19 Polyclonal antibody specifically detects ADAMTS19 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ADAMTS19
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Affinity Purified
ADAMTS19
This antibody was developed against a recombinant protein corresponding to amino acids: EPLNDTMAITGHPHRVYRQKRSMEEKVTEKSALHSHYCGIISDKGRPRSRKIAESGRGKRYSYKLPQEYNIETVVVADPAMVSYHGAD
Immunogen affinity purified
RUO
Primary
171019
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Q8TE59
A disintegrin and metalloproteinase with thrombospondin motifs 19, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 19, ADAM metallopeptidase with thrombospondin type 1 motif, 19, ADAM-TS 19, ADAM-TS19, ADAMTS-19, EC 3.4.24.-, FLJ16042
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only