Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA125179

Catalog No. PA125179


Add to cart

Description

Description

PA1-25179 detects ADAMTS2 from human samples. PA1-25179 has been successfully used in Western blot procedures. The PA1-25179 immunogen is a synthetic peptide corresponding to the C-terminal domain of human ADAMTS2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene excises the N-propeptide of type I, type II and type V procollagens. Mutations in this gene cause Ehlers-Danlos syndrome type VIIC, a recessively inherited connective-tissue disorder. Alternative splicing results in two transcript variants. The short transcript encodes a protein which has no significant procollagen N-peptidase activity.
Specifications

Specifications

ADAMTS2
Polyclonal
Unconjugated
ADAMTS2
Liquid
ADAMTS2
Synthetic peptide corresponding to the C-terminal domain of human ADAMTS2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.05% sodium azide
O95450
ADAM-TS2, ADAMTS-3, NPI, PCINP, PCPNI, hPCPNI
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
9509
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit