Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS4 Rabbit anti-Bovine, Human, Porcine, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA11750

Catalog No. PA11750


Add to cart

Description

Description

PA1-1750 detects ADAMTS4 from porcine, bovine, and human samples. PA1-1750 is specific for ADAMTS4 and shows no cross-reactivity with other ADAMTS family members. PA1-1750 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~68 and an~53 kDa protein representing ADAMTS4 from human recombit ADAMTS4. Depending on the extent of C-terminal processing, several smaller bands may also be seen using this antibody. PA1-1750 immunogen is a Synthetic peptide corresponding to residues C Y(590) N H R T D L F K S F P G P(603) of human ADAMTS4. This polyclonal antibody has been referred to as: JSCYNH.

A disintegrin-like and metalloproteinase thrombospondin type 1 motif 4 (ADAMTS4 or aggrecanase 1) is a member of the ADAMTS family of multidomain proteins that has demonstrated metalloproteinase functions. The ADAMTS4 protein contains a prohormone convertase cleavage site, thrombospondin-1 like domain, cysteine-rich domain, and a propeptide domain. ADAMTS4 activity had been shown to be blocked by TIMP-1, 2, 3, and 4. ADAMTS4 plays an important role in the cleavage of aggrecan, versican and brevican. Studies have determined that aggrecan is cleaved within the interglobular domain of aggrecan at sites Glu(373)-Ala(374) and Phe(342)-Glu(373). Aggrecan degradation has been linked to the development of rheumatoid arthritis, osteoarthritis, retinal diseases, Alzheimer’s and other neurodegenerative diseases.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

ADAMTS4
Polyclonal
Unconjugated
ADAMTS4
Liquid
ADAMTS4
Synthetic peptide corresponding to residues C Y(590) N H R T D L F K S F P G P(603) of human ADAMTS4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Porcine
Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q9TT93, O75173
Aggrecanase 1, A disintegrin and metalloproteinase with thrombospondin motifs 4, ADAM-TS 4, ADAM-TS4, ADMP-1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
100157276, 286806, 9507
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit