Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adaptin δ Mouse anti-Human, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611329

 View more versions of this product

Catalog No. BDB611329


Add to cart

Description

Description

Sorting of integral membrane proteins at various stages of the endocytic and secretory pathways is mediated by vesicular trafficking between a variety of organelles. Two sorting signals are tyrosine-based and dileucine-based signals that interact with heterotetrameric adaptor protein complexes (AP-1 through AP-4), which are associated with the vesicle coats. These coatomers contain two large adaptin proteins (γ, α, δ, ε and β1, β2, β3, β4, respectively) that are noncovalently linked to one medium chain (µ1, µ2, µ3, or µ4) and one small chain (σ1, σ2, σ3, or σ4). The AP-1 and AP-3 complexes are involved in protein sorting from the TGN and endosomes, while AP-2 adaptor complexes are involved in clathrin-mediated endocytosis. In the AP-3 complex, adaptin δ contains an N-terminal “head” domain that has 25% identity with adaptins α and γ, while its C-terminal “hinge” and “ear” domains are two-fold larger and more divergent as compared to the same domains in adaptins α and γ. Adaptin δ is widely expressed and its mutation results in protein storage deficiencies within lysosomal-related organelles in platelets, melansomes, and synaptic vesicles

Host Species: Mouse
Clone: 18
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Adaptin δ aa. 627-731
Formula Weight [Chemical]: 160kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

Adaptin δ
18
Unconjugated
Affinity Purified
Human Adaptin δ aa. 627-731
150μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.