Learn More
Adaptin ε Mouse anti-Human, Unlabeled, Clone: 32, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612018
Description
Sorting of integral membrane proteins is mediated by vesicular trafficking between a variety of organelles. Two sorting signals are tyrosine-based and dileucine-based signals that interact with heterotetrameric adaptor protein complexes (AP-1, AP-2, AP-3, and AP-4), which are associated with the vesicle coats. These coatomers contain two large adaptin proteins (γ, α, σ, ε and β1, β2, βσ2, σ3, σ4 respectively). The AP-1 and AP-3 complexes are involved in protein sorting from the TGN and endosomes, while AP-2 adaptor complexes are involved in clathrin-mediated endocytosis. AP-4 is associated with non-clathrin coated vesicles in the region of the TGN. This localization is disrupted by brefeldin A, indicating that AP-4 membrane attachment is regulated by small GTPases. The µ4 subunit of the AP-4 complex interacts with tyrosine-based signals on LAMP-2 during targeting to the endosomal-lysosomal system. Thus, AP-4 is a less abundant AP complex that may be important for vesicle trafficking from the Golgi to the endosomal-lysosomal system.
Specifications
Adaptin ε | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human Adaptin ε aa. 685-793 | |
50 μg | |
Cell Biology | |
Human | |
IgG1 |
Western Blot | |
32 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.